Learn More
Rainbow Scientific OPTIMAL HYPOTONIC SOLUTION
SDP
Catalog No. 501092828
Encompass
Developed for acute leukaemia bone marrow and tumour cells, also useful with problem blood and other problematic sample types.
Catalog No. 50-109-2828 Supplier Rainbow Scientific Supplier No. GGSJL005/A
Developed for acute leukaemia bone marrow and tumour cells, also useful with problem blood and other problematic sample types.