Learn More
Beckman Coulter OptiMATE Cesium Chloride, 500 mL
Catalog No. 502743745
Encompass
Highly concentrated, premixed CsCl solution offering significant time savings in reagent preparation. Ideal for continuous gradients requiring the highest quality purification, commonly used in viral, plasmid, or nucleic acid purification.Volume: 500mL, Density: 1.784 ± 0.014 g/mL, Concentration: 60% w/w*, Refractive Index: 1.408 ± 0.004, Endotoxin Level: ≤1 EU/mL, Sterile: Yes (autoclaved), Shelf Life: 18 months, Bottle Dimensions: 17.6cm x 7.2 cm / 6.93" x 2.83" (H x W)
Catalog No. 50-274-3745 Supplier Beckman Coulter Supplier No. D01357
Highly concentrated, premixed CsCl solution offering significant time savings in reagent preparation. Ideal for continuous gradients requiring the highest quality purification, commonly used in viral, plasmid, or nucleic acid purification.Volume: 500mL, Density: 1.784 ± 0.014 g/mL, Concentration: 60% w/w*, Refractive Index: 1.408 ± 0.004, Endotoxin Level: ≤1 EU/mL, Sterile: Yes (autoclaved), Shelf Life: 18 months, Bottle Dimensions: 17.6cm x 7.2 cm / 6.93" x 2.83" (H x W)