Learn More
Beckman Coulter OPTIMATE GRADIENT MAKER
Catalog No. NC3761907
Encompass
  • Automates pre-centrifugation steps including gradient preparation
  • Reduces manual intervention and potential errors
  • Enhances reproducibility across runs and operators
  • Reduces purification time by up to 75%
  • Reduces spin times from 48 hours to less than 5 hours
  • Compatible with various ultracentrifuge tubes
  • Ideal for purification of viral vectors and biomolecules
  • User-friendly interface for quick operator training
  • Compact design for flexible lab integration
  • Throughput of up to 6 tubes in 30–35 minutes
  • Supports liquid handling and scheduling software interface
  • Ambient temperature range of 10 to 35° C
  • Humidity range of 20% to 80% non-condensing
  • Compatible gradient media include CsCl, IDX, Sucrose
  • Gradient types of linear and step
Catalog No. NC3761907 Supplier Beckman Coulter Supplier No. D12917
Add to Cart
Add to Cart
Request a Quote
  • Automates pre-centrifugation steps including gradient preparation
  • Reduces manual intervention and potential errors
  • Enhances reproducibility across runs and operators
  • Reduces purification time by up to 75%
  • Reduces spin times from 48 hours to less than 5 hours
  • Compatible with various ultracentrifuge tubes
  • Ideal for purification of viral vectors and biomolecules
  • User-friendly interface for quick operator training
  • Compact design for flexible lab integration
  • Throughput of up to 6 tubes in 30–35 minutes
  • Supports liquid handling and scheduling software interface
  • Ambient temperature range of 10 to 35° C
  • Humidity range of 20% to 80% non-condensing
  • Compatible gradient media include CsCl, IDX, Sucrose
  • Gradient types of linear and step