Learn More
Beckman Coulter Beckman Coulter OptiMATE Lower Fluidics Cap for Gradient Maker (Conical/BEC)
Catalog No. NC3786532
Encompass
OptiMATE lower fluidics conical is a replacement lower fluidics cap assembly for the OptiMATE gradient maker. It includes a cap with an integrated dip tube and vent tubing with a filter to provide a secure fluid connection between gradient solution bottles or 250 mL conical sample containers and the instrument's upper fluidics path.
  • Includes cap with integrated dip tube and vent tubing with filter.
  • Connects CsCl and iodixanol gradient bottles and 250 mL conical sample containers to the upper fluidics path.
  • Quantity: 4 units.
  • Endotoxin: ≤ 10 EU/device.
  • Not autoclavable.
  • Designed to maintain sterile venting and reliable fluid transfer during gradient preparation.
Catalog No. NC3786532 Supplier Beckman Coulter Supplier No. D12220
OptiMATE lower fluidics conical is a replacement lower fluidics cap assembly for the OptiMATE gradient maker. It includes a cap with an integrated dip tube and vent tubing with a filter to provide a secure fluid connection between gradient solution bottles or 250 mL conical sample containers and the instrument's upper fluidics path.
  • Includes cap with integrated dip tube and vent tubing with filter.
  • Connects CsCl and iodixanol gradient bottles and 250 mL conical sample containers to the upper fluidics path.
  • Quantity: 4 units.
  • Endotoxin: ≤ 10 EU/device.
  • Not autoclavable.
  • Designed to maintain sterile venting and reliable fluid transfer during gradient preparation.