Learn More
Beckman Coulter Beckman Coulter OptiMATE Lower Fluidics (General) - Cap and tubing assembly for OptiMATE Gradient Maker
Catalog No. NC3786531 Shop All Beckman Coulter Products
Encompass
The OptiMATE Lower Fluidics (general) is a replacement lower-fluidics cap and tubing assembly for use with density gradient makers. It provides a secure cap, dip tube, and vent tubing with an inline filter to allow controlled reagent draw from general reagent bottles into the instrument's upper fluidics.
  • Includes cap with dip tube for reagent draw.
  • Includes vent tubing with integrated filter to reduce contamination risk.
  • Provides a secure connection between reagent bottles and the instrument fluidics.
  • Serves as a replacement accessory for routine maintenance and fluid management.
Catalog No. NC3786531 Supplier Beckman Coulter Supplier No. D12219
The OptiMATE Lower Fluidics (general) is a replacement lower-fluidics cap and tubing assembly for use with density gradient makers. It provides a secure cap, dip tube, and vent tubing with an inline filter to allow controlled reagent draw from general reagent bottles into the instrument's upper fluidics.
  • Includes cap with dip tube for reagent draw.
  • Includes vent tubing with integrated filter to reduce contamination risk.
  • Provides a secure connection between reagent bottles and the instrument fluidics.
  • Serves as a replacement accessory for routine maintenance and fluid management.