Learn More
Beckman Coulter Beckman Coulter OptiMATE Upper Fluidics - Replacement fluid path for OptiMATE Gradient Maker (D12216)
Catalog No. NC3786530
Encompass
The OptiMATE Upper Fluidics (D12216) is a consumable fluid path for the Beckman Coulter OptiMATE Gradient Maker that provides reagent pumping via peristaltic pumps, mixing in the manifold, and precise dispensing through the probe into sample tubes. It is supplied as a replaceable assembly to maintain consistent gradient preparation performance when used according to the instrument instructions for use.
  • Provides fluid routing for reagent pumping, mixing, and dispensing.
  • Includes two fluid assemblies per package.
  • Not autoclavable; intended as a disposable consumable.
  • Endotoxin level ≤ 10 EU per device.
  • Helps maintain consistent mixing and dispensing performance.
  • Refer to the instrument instructions for use for installation and handling.
Catalog No. NC3786530 Supplier Beckman Coulter Supplier No. D12216
The OptiMATE Upper Fluidics (D12216) is a consumable fluid path for the Beckman Coulter OptiMATE Gradient Maker that provides reagent pumping via peristaltic pumps, mixing in the manifold, and precise dispensing through the probe into sample tubes. It is supplied as a replaceable assembly to maintain consistent gradient preparation performance when used according to the instrument instructions for use.
  • Provides fluid routing for reagent pumping, mixing, and dispensing.
  • Includes two fluid assemblies per package.
  • Not autoclavable; intended as a disposable consumable.
  • Endotoxin level ≤ 10 EU per device.
  • Helps maintain consistent mixing and dispensing performance.
  • Refer to the instrument instructions for use for installation and handling.