Learn More
Optimize Technologies Optimize Universal Ferrule for 1/16" OD Tubing, PEEK
SDP
Catalog No. 50113418
Encompass_Preferred
OPTI-LOK™ Fittings are ideal for high-pressure connections with 1/16” OD tubing. All Optimize fittings are precision machined for optimal quality and thread consistency and are designed for trouble-free operation at pressures of up to 6,000 psi.
Catalog No. 50-113-418 Supplier Optimize Technologies Supplier No. 102400370
OPTI-LOK™ Fittings are ideal for high-pressure connections with 1/16” OD tubing. All Optimize fittings are precision machined for optimal quality and thread consistency and are designed for trouble-free operation at pressures of up to 6,000 psi.