Learn More
Optimize Technologies Optimize Universal Male Nut for 1/16" OD Tubing, 10-32, 316 Stainless Steel
SDP
Catalog No. 50113415
Encompass_Preferred
OPTI-LOK™ Fittings are ideal for high-pressure connections with 1/16” OD tubing. All Optimize fittings are precision machined for optimal quality and thread consistency and are designed for trouble-free operation at pressures of up to 6,000 psi.
Catalog No. 50-113-415 Supplier Optimize Technologies Supplier No. 102400367
OPTI-LOK™ Fittings are ideal for high-pressure connections with 1/16” OD tubing. All Optimize fittings are precision machined for optimal quality and thread consistency and are designed for trouble-free operation at pressures of up to 6,000 psi.