Learn More
Optimize Technologies Optimize Universal Stainless Steel 10-32 Nuts & SS Ferrules, 10pk (10-24-00367 & 10-24-00369)
SDP
Catalog No. 50113422
Encompass_Preferred
OPTI-LOK™ Fittings are ideal for high-pressure connections with 1/16” OD tubing. All Optimize fittings are precision machined for optimal quality and thread consistency and are designed for trouble-free operation at pressures of up to 6,000 psi.
Catalog No. 50-113-422 Supplier Optimize Technologies Supplier No. 102400376
OPTI-LOK™ Fittings are ideal for high-pressure connections with 1/16” OD tubing. All Optimize fittings are precision machined for optimal quality and thread consistency and are designed for trouble-free operation at pressures of up to 6,000 psi.