Learn More
Anton Paar Optimized for routine R&D and material formulationTorque range from 1 nNm to 215 mNmBroad range of temperature-based accessoriesQuickConnect coupling and Toolmaster featuresSuccessor to popular MCR 102e
Catalog No. 5000580467
Encompass_Preferred
MCR 303 SpecificationsBearing design: Air, fine-pored carbonMotor design: Electronically commutated (EC) permanent magnet synchronous motorDisplacement transducer: High-resolution optical encoderNormal force measurement design: 360° capacitive sensor, non-contacting, fully integrated in bearingWorking Modules: CMTMin. torque (rotation): 5 nNmMin. torque (oscillation): 5/1 nNmMax. torque: 215 mNmTorque resolution: 0.1 nNmAngular deflection resolution: 3 nradMin. angular velocity: 0 rad/sMax. angular velocity / max. speed: 314 rad/s (3,000 rpm)Min. frequency: 2 x 10-8 HzMax. frequency: 100 HzNormal force range: 0.001 N to 50 NNormal force resolution: 0.1 mNTruStrain: OptionalReady for DMA in tension, bending, and compression: n/a
Catalog No. 50-005-80467 Supplier Anton Paar Supplier No. 220247
Please contact your sales representative for a quote or further ordering instructions.
MCR 303 SpecificationsBearing design: Air, fine-pored carbonMotor design: Electronically commutated (EC) permanent magnet synchronous motorDisplacement transducer: High-resolution optical encoderNormal force measurement design: 360° capacitive sensor, non-contacting, fully integrated in bearingWorking Modules: CMTMin. torque (rotation): 5 nNmMin. torque (oscillation): 5/1 nNmMax. torque: 215 mNmTorque resolution: 0.1 nNmAngular deflection resolution: 3 nradMin. angular velocity: 0 rad/sMax. angular velocity / max. speed: 314 rad/s (3,000 rpm)Min. frequency: 2 x 10-8 HzMax. frequency: 100 HzNormal force range: 0.001 N to 50 NNormal force resolution: 0.1 mNTruStrain: OptionalReady for DMA in tension, bending, and compression: n/a