Learn More
Cambridge Isotope Laboratories OPTIMIZER INSERT FOR ADAPTING 3MM NMR TUBE TO 5MM BRUKER SPINNERS, EA
Catalog No. 501629050
Encompass
OPTIMIZER INSERT FOR ADAPTING 3MM NMR TUBE TO 5MM BRUKER SPINNERS, EA
Catalog No. 50-162-9050 Supplier Cambridge Isotope Laboratories Supplier No. 5X3INSB
OPTIMIZER INSERT FOR ADAPTING 3MM NMR TUBE TO 5MM BRUKER SPINNERS, EA