Learn More
Sigma Aldrich Fine Chemicals Biosciences Optimizer insert for spinn1EA
Catalog No. 502901078
Encompass
Precision adapter holds 3 mm 4 mm and 4.25 mm NMR tube in a 5 mm spinner turbine. Does not include spinner turbine.
Catalog No. 50-290-1078 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. NOR5X3INSB1EA
Precision adapter holds 3 mm 4 mm and 4.25 mm NMR tube in a 5 mm spinner turbine. Does not include spinner turbine.