Learn More
ELITechGroup OPTIMOLE™ Osmolality Standard, 290 mmol/kg, 60x0.4mL
SDP
Catalog No. NC9306976
Encompass
.4MLx60, Standard, Osmolality; Opti-Mole; Wescor; Sodium Chloride Solution, Saline; Liquid, clear odorless; B.P. 100 deg.C; Freezing Point 0 deg.C; Storage: RT away from direct light; 290Mmol/kg
Catalog No. NC9306976 Supplier ELITechGroup Supplier No. OA029
.4MLx60, Standard, Osmolality; Opti-Mole; Wescor; Sodium Chloride Solution, Saline; Liquid, clear odorless; B.P. 100 deg.C; Freezing Point 0 deg.C; Storage: RT away from direct light; 290Mmol/kg