Learn More
ELITechGroup OPTIMOLE™ Osmolality Standard, 1000 mmol/kg, 60x0.4mL
SDP
Catalog No. NC9306975
Encompass
Standard, Osmolality; Opti-Mole; Wescor; Sodium Chloride Solution, Saline; Liquid, clear odorless; B.P. 100 deg.C; Freezing Point 0 deg.C; Storage: RT away from direct light; 1000Mmol/kg
Catalog No. NC9306975 Supplier ELITechGroup Supplier No. OA100
Standard, Osmolality; Opti-Mole; Wescor; Sodium Chloride Solution, Saline; Liquid, clear odorless; B.P. 100 deg.C; Freezing Point 0 deg.C; Storage: RT away from direct light; 1000Mmol/kg