Learn More
Thomson Instrument Company Optimum Growth 125mL 8 Position Carrier
SDP
Catalog No. 502221318
Encompass_Preferred
Designed for use with the Optimum Growth 125 mL Flask (P/N: 931110), this sturdy carrier allows for the easy and efficient transport of up to 8 flasks at once. Ideal for streamlining workflow in lab environments, it ensures secure handling while minimizing the risk of spills or breakage during transfer. Made in the USA
Catalog No. 50-222-1318 Supplier Thomson Instrument Company Supplier No. 1212900
Designed for use with the Optimum Growth 125 mL Flask (P/N: 931110), this sturdy carrier allows for the easy and efficient transport of up to 8 flasks at once. Ideal for streamlining workflow in lab environments, it ensures secure handling while minimizing the risk of spills or breakage during transfer. Made in the USA