Learn More
Thomson Instrument Company Optimum Growth 250mL Flask, w/ Sampling Port and Vent Cap, Sterile
SDP
Catalog No. NC1375983
Encompass_Preferred
Thomson’s Optimum Growth Sampling Flasks feature integrated one-way valves that enable aseptic sampling directly in the shaker or on the benchtop, helping reduce contamination risk and improve efficiency—especially in high-throughput environments.
•Available in 125 mL, 250 mL, 500 mL, and 5 L sizes with integrated one-way sampling valves
•Aseptic sampling without removing the cap—simply connect a syringe and collect samples
•Enables sampling without removing flasks from shakers or moving them to biosafety cabinets
•Streamlines workflows and minimizes time, contamination risk, and handling complexity
•0.2 µm PTFE vented caps support sterile gas exchange and maintain culture integrity
• Made in the USA
Catalog No. NC1375983 Supplier Thomson Instrument Company Supplier No. 931111SP
Thomson’s Optimum Growth Sampling Flasks feature integrated one-way valves that enable aseptic sampling directly in the shaker or on the benchtop, helping reduce contamination risk and improve efficiency—especially in high-throughput environments.
•Available in 125 mL, 250 mL, 500 mL, and 5 L sizes with integrated one-way sampling valves
•Aseptic sampling without removing the cap—simply connect a syringe and collect samples
•Enables sampling without removing flasks from shakers or moving them to biosafety cabinets
•Streamlines workflows and minimizes time, contamination risk, and handling complexity
•0.2 µm PTFE vented caps support sterile gas exchange and maintain culture integrity
• Made in the USA