Learn More
Thomson Instrument Company Optimum Growth Solid Cap, Sterile
SDP
Catalog No. NC2941118
Encompass_Preferred
Designed for use with Optimum Growth 1.6 L, 2.8 L, and 5 L Flasks, this sterile storage cap features a low-evaporation design to minimize media loss during storage and incubation. Ideal for long-term or static culture conditions, it ensures a secure seal while maintaining product integrity.Flask Compatibility: 1.6L, 2.8L, 5LSterility (SAL): 10⁻⁶ Made in the USA
Catalog No. NC2941118 Supplier Thomson Instrument Company Supplier No. 899600B
Designed for use with Optimum Growth 1.6 L, 2.8 L, and 5 L Flasks, this sterile storage cap features a low-evaporation design to minimize media loss during storage and incubation. Ideal for long-term or static culture conditions, it ensures a secure seal while maintaining product integrity.Flask Compatibility: 1.6L, 2.8L, 5LSterility (SAL): 10⁻⁶ Made in the USA