Learn More
Cayman Chemical OptInurInC-Term Blockng 1ea
SDP
Catalog No. 502988775
Encompass_Preferred
A long-chain atypical ceramide contains a sphingosine (d181(14Z)) backbone with a cis double bond at the 14-15 position
Catalog No. 50-298-8775 Supplier Cayman Chemical Supplier No. 3000001 EA
A long-chain atypical ceramide contains a sphingosine (d181(14Z)) backbone with a cis double bond at the 14-15 position