Learn More
Cayman Chemical OptInurInC-Term Monclona 100ug
SDP
Catalog No. 502938857
Encompass_Preferred
Immunogen: Peptide from C-Terminal region of human optineurin protein • Host: Mouse • Species Reactivity: (+) Human • Applications: Immunocytochemistry • MW = 66 kDa
Catalog No. 50-293-8857 Supplier Cayman Chemical Supplier No. 27230100 UG
Immunogen: Peptide from C-Terminal region of human optineurin protein • Host: Mouse • Species Reactivity: (+) Human • Applications: Immunocytochemistry • MW = 66 kDa