Learn More
Molecular Devices Molecular Devices Reservoir, 4 L, Without Level Sensor
Catalog No. 5000014691
Encompass_Preferred
4 L reservoir designed for reagent or waste containment in microplate washer workflows. This replacement bottle does not include an integrated level sensor and is suitable for use with compatible AquaMax 2000 and 4000 washers.
  • 4 L capacity.
  • No integrated level sensor.
  • Designed for reagent or waste containment during plate washing.
  • Replacement bottle for compatible microplate washers.
Catalog No. 50-000-14691 Supplier Molecular Devices Supplier No. 5315485
Add to Cart
Add to Cart
Request a Quote
4 L reservoir designed for reagent or waste containment in microplate washer workflows. This replacement bottle does not include an integrated level sensor and is suitable for use with compatible AquaMax 2000 and 4000 washers.
  • 4 L capacity.
  • No integrated level sensor.
  • Designed for reagent or waste containment during plate washing.
  • Replacement bottle for compatible microplate washers.