Learn More
Research Products International Corp Optional 3-D Orbital Tray for Roto-Shake Genie
SDP
Catalog No. 501977230
Encompass
The 3-D Orbital Shaker Attachment for the Roto-Shake Genie provides gentle yet thorough mixing of dishes, flasks, beakers and open containers. It moves at a 5 degree angle through a 360 degree rotation (providing a combination of vertical and horizontal motions). This means that every corner or crevice of a container will receive complete mixing, making it the perfect choice for applications such as gel and blot staining/blocking, sample washing, polymer resin preparation, and more.
Please note, platform capacity is 1 Kg (2.2 lb).
Catalog No. 50-197-7230 Supplier Research Products International Corp Supplier No. 2488163D
The 3-D Orbital Shaker Attachment for the Roto-Shake Genie provides gentle yet thorough mixing of dishes, flasks, beakers and open containers. It moves at a 5 degree angle through a 360 degree rotation (providing a combination of vertical and horizontal motions). This means that every corner or crevice of a container will receive complete mixing, making it the perfect choice for applications such as gel and blot staining/blocking, sample washing, polymer resin preparation, and more.
Please note, platform capacity is 1 Kg (2.2 lb).