Learn More
Chemglass Life Sciences Optional CO2 Gas Regulator for SureTherm CO2 Incubators
SDP
Catalog No. 5000083271
Encompass
Optional CO2 Gas Regulator provides the required connection and regulation of CO2 from an external cylinder to SureTherm CO2 incubators. It enables controlled CO2 delivery for cell culture applications when an external gas source is used.
  • Intended for connecting an external CO2 cylinder to a CO2 incubator.
  • Regulates CO2 supply to maintain a controlled atmosphere for cell culture.
  • Supplies an optional external gas connection when an internal source is not used.
  • Compact accessory designed for easy installation alongside incubator gas inlets.
Catalog No. 50-000-83271 Supplier Chemglass Life Sciences Supplier No. CLS2460REG
Optional CO2 Gas Regulator provides the required connection and regulation of CO2 from an external cylinder to SureTherm CO2 incubators. It enables controlled CO2 delivery for cell culture applications when an external gas source is used.
  • Intended for connecting an external CO2 cylinder to a CO2 incubator.
  • Regulates CO2 supply to maintain a controlled atmosphere for cell culture.
  • Supplies an optional external gas connection when an internal source is not used.
  • Compact accessory designed for easy installation alongside incubator gas inlets.