Learn More
World Precision Instrument Optional Foot Switch for Micro2T, Micro-ePore™, DMF1000
SDP
Catalog No. NC1485369
Encompass
  • MF200-FS is for the DMF1000 and MF200
  • 13142 is for the MICRO2T, MICRO-ePORE and EVOM3
Catalog No. NC1485369 Supplier World Precision Instrument Supplier No. 13142
  • MF200-FS is for the DMF1000 and MF200
  • 13142 is for the MICRO2T, MICRO-ePORE and EVOM3