Learn More
Alkali Scientific Horizontal head, for 2 x 50mL for BenchMixer™ (BV1000 & BV1003)
SDP
Catalog No. 502389308
Encompass_Preferred
Optional horizontal head, for 2 x 50mL for the Benchmark Mortexer Vortex Mixer RS7001. Holds two 50mL tubes.
Catalog No. 50-238-9308 Supplier Alkali Scientific Supplier No. RS7001H500
Optional horizontal head, for 2 x 50mL for the Benchmark Mortexer Vortex Mixer RS7001. Holds two 50mL tubes.