Learn More
Bellco Glass, Inc. Optional Temperature Alarm Roll-in Incubators
SDP
Catalog No. 501946061
Encompass_Preferred
****FACTORY INSTALLED********NOT FOR REFRIGERATED UNITS****Bellco optional temperature alarm module is designed for use with standard 7728 series roll-in incubators including the mini roll-inc.(not refri units). In operation, the alarm module monitors the alarm output of the temperature controller. The alarm value may be set to temperatures between 0-50°c. If the internal temperature of the incubator exceeds the alarm value, then an audible warning is produced. In addition, an internal relay is closed, activating any alarm circuitry connected to the remote jack.. MADE IN THE USA
Catalog No. 50-194-6061 Supplier Bellco Glass, Inc. Supplier No. 772880000
****FACTORY INSTALLED********NOT FOR REFRIGERATED UNITS****Bellco optional temperature alarm module is designed for use with standard 7728 series roll-in incubators including the mini roll-inc.(not refri units). In operation, the alarm module monitors the alarm output of the temperature controller. The alarm value may be set to temperatures between 0-50°c. If the internal temperature of the incubator exceeds the alarm value, then an audible warning is produced. In addition, an internal relay is closed, activating any alarm circuitry connected to the remote jack.. MADE IN THE USA