Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Set of Two Heratrays

Catalog No. 11688013 Shop All Thermo Scientific Products
Change view
Click to view available options
Material:
Copper
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
11-688-013 Copper
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-688-013 Supplier Thermo Scientific™ Supplier No. 50061050
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Material: Copper; For Use With: Heracell 150i

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Heracell 150i
Lighting LED
Material Copper
Type Tray
Width (English) 0.5 in.
Width (Metric) 1.27 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.