Learn More
Cosmo Bio Usa Inc OptiPrep™ 1x250ml
SDP
Catalog No. NC1059560
Encompass
OptiPrep™ is a sterile and endotoxin tested multi-purpose medium based on iodixanol. Unlike any of these other gradient media, iodixanol (as an X-ray imaging agent) has been clinically tested and is manufactured under strict and EU cGMP compliance.OptiPrep™ specifications:Iodixanol: 60% (w/v)Density: 1.320 ± 0.001 g/mlOsmolality: approx. 170 mOsmEndotoxins: < 1.0 EU/ml
Catalog No. NC1059560 Supplier Cosmo Bio Usa Inc Supplier No. AXS1114542
OptiPrep™ is a sterile and endotoxin tested multi-purpose medium based on iodixanol. Unlike any of these other gradient media, iodixanol (as an X-ray imaging agent) has been clinically tested and is manufactured under strict and EU cGMP compliance.OptiPrep™ specifications:Iodixanol: 60% (w/v)Density: 1.320 ± 0.001 g/mlOsmolality: approx. 170 mOsmEndotoxins: < 1.0 EU/ml