Learn More
STEMCELL Technologies OptiPrep™, Size: 250 mL
SDP
Catalog No. NC1339550
Encompass
Simplify the isolation and purification of macromolecules; viruses; a wide range of cell types; and organelles such as nuclei, mitochondria, endosomes, or exosomes by using OptiPrep™ medium. This flexible, versatile, and gentle density gradient medium is non-ionic, iodixanol-based (60% w/v) and has a density of 1.320 ± 0.001 g/mL. OptiPrep™ has been used clinically in X-ray contrast imaging and has been shown to have lower toxicity when compared to ionic gradient compounds such as metrizamide.
Catalog No. NC1339550 Supplier STEMCELL Technologies Supplier No. 07820
Simplify the isolation and purification of macromolecules; viruses; a wide range of cell types; and organelles such as nuclei, mitochondria, endosomes, or exosomes by using OptiPrep™ medium. This flexible, versatile, and gentle density gradient medium is non-ionic, iodixanol-based (60% w/v) and has a density of 1.320 ± 0.001 g/mL. OptiPrep™ has been used clinically in X-ray contrast imaging and has been shown to have lower toxicity when compared to ionic gradient compounds such as metrizamide.