Learn More
Hach Company OptiQuant SST Calibration Kit for OptiQuant T-Line and Immersion Probes
Catalog No. NC3641060 Shop All Hach Company Products
Encompass
The OptiQuant SST Calibration Kit from Hach provides the standards and accessories required to calibrate SST/turbidity probes used with OptiQuant T-Line systems. The kit supplies a stable high-range turbidity standard and the hardware needed for consistent calibrations in process and field environments.
  • Includes a calibration cylinder for probe alignment.
  • Contains two 500 mL bottles of 800 NTU turbidity standard.
  • Includes a probe bracket for immersion turbidity probes.
  • Designed to enable accurate and repeatable turbidity calibrations.
  • Suitable for use in online and field measurement setups.
Catalog No. NC3641060 Supplier Hach Company Supplier No. 5733000
The OptiQuant SST Calibration Kit from Hach provides the standards and accessories required to calibrate SST/turbidity probes used with OptiQuant T-Line systems. The kit supplies a stable high-range turbidity standard and the hardware needed for consistent calibrations in process and field environments.
  • Includes a calibration cylinder for probe alignment.
  • Contains two 500 mL bottles of 800 NTU turbidity standard.
  • Includes a probe bracket for immersion turbidity probes.
  • Designed to enable accurate and repeatable turbidity calibrations.
  • Suitable for use in online and field measurement setups.