Learn More
EMD MILLIPORE 3P BIOPHARM OPTISCALE FILTER EXPRESS 3/PK
Supplier:  EMD MILLIPORE 3P BIOPHARM SGEPA47HH3
Encompass
Optiscale 47mm Millipore Express SHF 0.2um 1/4in. HB/HB
Catalog No. NC0292524