Learn More
Beckman Coulter Beckman Coulter OptiSeal Tube Kit, 32.4 mL, 25 x 77 mm (MPN 361662)
Catalog No. NC1987484
Encompass_Preferred
The OptiSeal Tube Kit provides a complete set of sealed ultracentrifuge tubes, spacers, and tools for OptiSeal-compatible rotors. It supplies tubes and hardware to support secure sealing, extraction, and organized storage during ultracentrifugation.
  • Includes four boxes of sealed ultracentrifuge tubes (224 total tubes).
  • Tubes are polyallomer, 32.4 mL, 25 x 77 mm.
  • Includes 12 spacers (six packages of two).
  • Includes one tube rack for organized storage.
  • Includes a tube extraction tool and a spacer removal tool.
  • Provides components for OptiSeal-compatible rotor workflows.
Catalog No. NC1987484 Supplier Beckman Coulter Supplier No. 361662
The OptiSeal Tube Kit provides a complete set of sealed ultracentrifuge tubes, spacers, and tools for OptiSeal-compatible rotors. It supplies tubes and hardware to support secure sealing, extraction, and organized storage during ultracentrifugation.
  • Includes four boxes of sealed ultracentrifuge tubes (224 total tubes).
  • Tubes are polyallomer, 32.4 mL, 25 x 77 mm.
  • Includes 12 spacers (six packages of two).
  • Includes one tube rack for organized storage.
  • Includes a tube extraction tool and a spacer removal tool.
  • Provides components for OptiSeal-compatible rotor workflows.