Learn More
Beckman Coulter OptiSeal Tube Rack for 25 x 89 mm (1 x 3.5 in) ultracentrifuge tubes
Catalog No. NC2856091
Encompass_Preferred
The OptiSeal Tube Rack is an accessory designed to hold and organize sealed ultracentrifuge tubes measuring 25 x 89 mm (1 x 3.5 in). It provides secure storage and makes handling sealed tubes easier during preparation and storage.
  • Holds sealing ultracentrifuge tubes measuring 25 x 89 mm (1 x 3.5 in).
  • Typical nominal tube volume: 36.2 mL.
  • Used for organization and secure storage of sealed centrifuge tubes.
  • Designed as a rack assembly for sealed tube handling and transport.
Catalog No. NC2856091 Supplier Beckman Coulter Supplier No. 360542
The OptiSeal Tube Rack is an accessory designed to hold and organize sealed ultracentrifuge tubes measuring 25 x 89 mm (1 x 3.5 in). It provides secure storage and makes handling sealed tubes easier during preparation and storage.
  • Holds sealing ultracentrifuge tubes measuring 25 x 89 mm (1 x 3.5 in).
  • Typical nominal tube volume: 36.2 mL.
  • Used for organization and secure storage of sealed centrifuge tubes.
  • Designed as a rack assembly for sealed tube handling and transport.