Learn More
Beckman Coulter OPTISEAL TUBES 5/8X2-3/4 56/BX
Catalog No. NC9611575
Encompass_Preferred
Polyallomer OptiSeal Tubes,Quantity:56,Size(mm):16 x 70, Vol.(mL): 11.2
Catalog No. NC9611575 Supplier Beckman Coulter Supplier No. 362181
Polyallomer OptiSeal Tubes,Quantity:56,Size(mm):16 x 70, Vol.(mL): 11.2