Learn More
Opti Temp OPTISHIELD II CORROSION 1 GALL
SDP
Catalog No. NC1379094
Encompass
OptiShield II Low Conductivity Formula OptiShield II corrosion inhibitor has been specially formulated to provide corrosion protection in systems such as laser, welding, induction, and other equipment where "electrical conductivity" of the fluid is of concern
Catalog No. NC1379094 Supplier Opti Temp Supplier No. 400108
OptiShield II Low Conductivity Formula OptiShield II corrosion inhibitor has been specially formulated to provide corrosion protection in systems such as laser, welding, induction, and other equipment where "electrical conductivity" of the fluid is of concern