Learn More
Electron Microscopy Sciences Optivison™ Optivisor Add-On Light
SDP
Catalog No. 5030521
Encompass_Preferred
Features a dial adjustment headband that fits any head size. Wellmade magnifiers they are lightweight and durable, and fit over eyeglasses. Adjustable pivot assembly on visor can be set to desired tension, so it will stay in place when tilted. Lenses are ground and polished prismatic. Lens plate replacement is also available. Add on light for Optivisor
Catalog No. 50-305-21 Supplier Electron Microscopy Sciences Supplier No. 6810760
Features a dial adjustment headband that fits any head size. Wellmade magnifiers they are lightweight and durable, and fit over eyeglasses. Adjustable pivot assembly on visor can be set to desired tension, so it will stay in place when tilted. Lenses are ground and polished prismatic. Lens plate replacement is also available. Add on light for Optivisor