Learn More
Med Vet International Optivisor Replacement Lens Plate, 1.5x Magnification, 20", J0801LPA
Catalog No. 5000583824
Encompass_Preferred
Only for optivisor. These lenses interchange by removing two screws.
Catalog No. 50-005-83824 Supplier Med Vet International Supplier No. JOR801LPA
Only for optivisor. These lenses interchange by removing two screws.