Learn More
Med Vet International Optivisor Replacement Lens Plate 2x Magnification, 10", J0801LPC
Catalog No. 5000583863
Encompass_Preferred
Only for optivisor. These lenses interchange by removing two screws.
Catalog No. 50-005-83863 Supplier Med Vet International Supplier No. JOR801LPC
Only for optivisor. These lenses interchange by removing two screws.