Learn More
Beckman Coulter OptiXTRACT Sample Recovery Aid (D17685) - density gradient band extraction aid
Catalog No. NC3920953
Encompass_Preferred
Beckman Coulter's OptiXTRACT Sample Recovery Aid provides clarity and control for density gradient band extraction. The device's tube mount, guided syringe rail, LED illumination, and millimeter graduations help users visualize bands and perform accurate, consistent, and reproducible recovery of multiple sample bands from centrifuge tubes.
  • Tube mount for secure positioning of tubes.
  • Guided syringe rail for controlled sample extraction.
  • LED illumination for clear band visualization.
  • Millimeter graduations for reproducible sampling.
  • Enables accurate and consistent recovery of multiple bands.
  • Designed for density gradient band extraction with centrifuge tubes.
Catalog No. NC3920953 Supplier Beckman Coulter Supplier No. D17685
Beckman Coulter's OptiXTRACT Sample Recovery Aid provides clarity and control for density gradient band extraction. The device's tube mount, guided syringe rail, LED illumination, and millimeter graduations help users visualize bands and perform accurate, consistent, and reproducible recovery of multiple sample bands from centrifuge tubes.
  • Tube mount for secure positioning of tubes.
  • Guided syringe rail for controlled sample extraction.
  • LED illumination for clear band visualization.
  • Millimeter graduations for reproducible sampling.
  • Enables accurate and consistent recovery of multiple bands.
  • Designed for density gradient band extraction with centrifuge tubes.