Learn More
Aviva Systems Biology Corp OPTN Antibody - N-terminal region (ARP39525_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39525_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OPTN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human OPTN. Alternate names: FIP2, GLC1E, HIP7, HYPL, NRP, TFIIIA-INTP, ALS12ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rat
Catalog No. 50-014-57073