Learn More
BRUKER CELLULAR ANALYSIS INC OPTO SELECT 1750B CHIP
SDP
Catalog No. NC2410762
Encompass
  • Disposable single-use nanofluidic chip
  • Compatible with Beacon Optofluidic Systems
  • Compatible with Beacon Select Optofluidic Systems
  • Contains approximately 1,750 NanoPen chambers
  • Optimized for on-chip cell culturing
  • Intended for cell line development workflows
  • Must be stored at room temperature
  • Wetting solution should be stored at 4°C
  • Minimum shelf life of 6 months
  • Use powder-free nitrile gloves during handling
  • Recommended cleaning with 99% isopropyl alcohol (IPA)
  • Features large pen geometry for effective cell culture
Catalog No. NC2410762 Supplier BRUKER CELLULAR ANALYSIS INC Supplier No. 75000018
  • Disposable single-use nanofluidic chip
  • Compatible with Beacon Optofluidic Systems
  • Compatible with Beacon Select Optofluidic Systems
  • Contains approximately 1,750 NanoPen chambers
  • Optimized for on-chip cell culturing
  • Intended for cell line development workflows
  • Must be stored at room temperature
  • Wetting solution should be stored at 4°C
  • Minimum shelf life of 6 months
  • Use powder-free nitrile gloves during handling
  • Recommended cleaning with 99% isopropyl alcohol (IPA)
  • Features large pen geometry for effective cell culture