Learn More
Electron Microscopy Sciences Opto-Wipes™ Size 4x4"
SDP
Catalog No. 50949296
Encompass_Preferred
A strong hydro-entangled mixture of polyester and cellulose fiber; designed for clean room environments. This special material offers softness, is lint-free, and traps particles and residue quite easily.
Soft: super-soft fibers to prevent the scratching of precision surfaces.
Durable: Strong fiber matrix maintains integrity when wet resulting in durability.
Highly absorbent by retaining and withstanding solvents such as MEK, methylene chloride, acetone, and alcohol.
Particle trapping matrix lifts particles and residue away from the surface. Washable and reusable.
Catalog No. 50-949-296 Supplier Electron Microscopy Sciences Supplier No. 7175501
A strong hydro-entangled mixture of polyester and cellulose fiber; designed for clean room environments. This special material offers softness, is lint-free, and traps particles and residue quite easily.
Soft: super-soft fibers to prevent the scratching of precision surfaces.
Durable: Strong fiber matrix maintains integrity when wet resulting in durability.
Highly absorbent by retaining and withstanding solvents such as MEK, methylene chloride, acetone, and alcohol.
Particle trapping matrix lifts particles and residue away from the surface. Washable and reusable.