Learn More
Electron Microscopy Sciences Opto-Wipes™ Size 6x12"
SDP
Catalog No. 50949348
Encompass_Preferred
A strong hydro-entangled mixture of polyester and cellulose fiber; designed for clean room environments. This special material offers softness, is lint-free, and traps particles and residue quite easily.
Soft: super-soft fibers to prevent the scratching of precision surfaces.
Durable: Strong fiber matrix maintains integrity when wet resulting in durability.
Highly absorbent by retaining and withstanding solvents such as MEK, methylene chloride, acetone, and alcohol.
Particle trapping matrix lifts particles and residue away from the surface. Washable and reusable.
This item is not returnable. View return policy
Catalog No. 50-949-348 Supplier Electron Microscopy Sciences Supplier No. 7175503
A strong hydro-entangled mixture of polyester and cellulose fiber; designed for clean room environments. This special material offers softness, is lint-free, and traps particles and residue quite easily.
Soft: super-soft fibers to prevent the scratching of precision surfaces.
Durable: Strong fiber matrix maintains integrity when wet resulting in durability.
Highly absorbent by retaining and withstanding solvents such as MEK, methylene chloride, acetone, and alcohol.
Particle trapping matrix lifts particles and residue away from the surface. Washable and reusable.