Learn More
Medchemexpress LLC HY-12809 5mg Medchemexpress, Optovin CAS:348575-88-2 Purity:>98%
SDP
Catalog No. 502081149
Encompass_Preferred
Medchemexpress, HY-12809 5mg Optovin CAS:348575-88-2 Optovin is a reversible photoactivated TRPA1 ligand that enables light-mediated neuronal excitation. Optovin activates TRPA1 via structure-dependent photochemical reactions with redox-sensitive cysteine residues. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1149 Supplier Medchemexpress LLC Supplier No. HY128095MG
Medchemexpress, HY-12809 5mg Optovin CAS:348575-88-2 Optovin is a reversible photoactivated TRPA1 ligand that enables light-mediated neuronal excitation. Optovin activates TRPA1 via structure-dependent photochemical reactions with redox-sensitive cysteine residues. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.