Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Optovin, Tocris Bioscience™

Catalog No. 490150
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-015-0 50 mg
49-011-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 49-015-0 Supplier R&D Systems Supplier No. 4901/50
This item is not returnable. View return policy

Reversible photoactive TRPA1 activator

Reversible photoactive TRPA1 activator. Stimulates human TRPA1 channels in vitro and enables repeated photoactivation of motor behaviors in wild-type zebrafish (EC50 = 2 μM) and mice, in vivo.

Chemical Identifiers

CAS 348575-88-2
Molecular Formula C15H13N3OS2
Molecular Weight (g/mol) 315.409
InChI Key YISGMOZSGOGYOU-UHFFFAOYSA-N
Synonym optovin, cbmicro_018200, 5e-5-2,5-dimethyl-1-pyridin-3-ylpyrrol-3-yl methylidene-2-sulfanylidene-1,3-thiazolidin-4-one, 5-2,5-dimethyl-1-pyridin-3-ylpyrrol-3-yl methylidene-2-sulfanylidene-1,3-thiazolidin-4-one
PubChem CID 2890284
IUPAC Name 5-[(2,5-dimethyl-1-pyridin-3-ylpyrrol-3-yl)methylidene]-2-sulfanylidene-1,3-thiazolidin-4-one
SMILES CC1=CC(=C(N1C2=CN=CC=C2)C)C=C3C(=O)NC(=S)S3

Specifications

Quantity 50 mg
Formula Weight 315.41
Percent Purity >99%
Chemical Name or Material Optovin
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.