Learn More
Thermo Scientific Caffeine/Pyrene Standard Kit 1EA
Supplier:  Thermo Scientific 3323.0010
Encompass
Caffeine and pyrene HPLC standard reference material for quantitative analysis and calibration standard containing precisely weighed amounts of caffeine and pyrene dissolved in inert matrix.
Catalog No. 50-134-6987