Learn More
Medchemexpress LLC Or-1896 | 220246-81-1 | MFCD09952125 | 99.0% | 245.28 g/mol | C13H15N3O2 | 5 MG
SDP
Catalog No. 5000222152
Encompass_Preferred
OR-1896 is the active, long-lived metabolite of levosimendan. It functions as a calcium sensitizer and a selective phosphodiesterase III inhibitor, and can open ATP-sensitive potassium channels producing vasodilatory effects. The compound is supplied for research use as a solid powder and as a 10 mM solution in DMSO.
  • Calcium sensitizer and phosphodiesterase III inhibitor.
  • Opens ATP-sensitive potassium channels and produces vasodilation.
  • Suitable for in vitro pharmacology and cardiomyocyte studies.
  • Available as powder in small-scale sizes and as a DMSO solution.
  • High reported purity (~99%) appropriate for research use.
  • Accompanied by safety data sheet and standard storage recommendations.
Catalog No. 50-002-22152 Supplier Medchemexpress LLC Supplier No. HY1357465MG
OR-1896 is the active, long-lived metabolite of levosimendan. It functions as a calcium sensitizer and a selective phosphodiesterase III inhibitor, and can open ATP-sensitive potassium channels producing vasodilatory effects. The compound is supplied for research use as a solid powder and as a 10 mM solution in DMSO.
  • Calcium sensitizer and phosphodiesterase III inhibitor.
  • Opens ATP-sensitive potassium channels and produces vasodilation.
  • Suitable for in vitro pharmacology and cardiomyocyte studies.
  • Available as powder in small-scale sizes and as a DMSO solution.
  • High reported purity (~99%) appropriate for research use.
  • Accompanied by safety data sheet and standard storage recommendations.