Learn More
World Precision Instrument Sensor 200U NO, 2PK Custom L
SDP
Catalog No. 502521159
Encompass
ISO-NOPF electrodes are available in 100 µm, 200 µm and 500 µm diameters. Utilizing the latest advances in nano-technology and material science, scientists at WPI’s Sensor Laboratory have created these completely flexible and virtually unbreakable NO sensors. The new sensors are based on a composite graphite NO-sensing element combined with a reference electrode. The surface of the sensor is then coated with a unique multi-layered NO-selective membrane.
Catalog No. 50-252-1159 Supplier World Precision Instrument Supplier No. ISONOPF200LXX
ISO-NOPF electrodes are available in 100 µm, 200 µm and 500 µm diameters. Utilizing the latest advances in nano-technology and material science, scientists at WPI’s Sensor Laboratory have created these completely flexible and virtually unbreakable NO sensors. The new sensors are based on a composite graphite NO-sensing element combined with a reference electrode. The surface of the sensor is then coated with a unique multi-layered NO-selective membrane.