Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surgical Design™ OR Grade Hartman Mosquito Hemostatic Forceps, Anti-glare

Catalog No. 21124983 Shop All Surgical Design Products
Catalog No. 21-124-983 Supplier Surgical Design™ Supplier No. ORG159S
Add to Cart
Add to Cart

O.R.-grade Hartman mosquito forceps with anti-glare finish are designed for precise clamping in delicate surgical fields.

  • Length: 3.5 in.
  • Tip Options: Straight or Curved
  • O.R. Grade construction with an anti-glare satin finish to reduce reflections during procedures
  • Fine, narrow jaws ideal for delicate tissue handling and small-vessel hemostasis
  • Precision serrations provide secure, non-slip grip
  • Smooth ratchet lock ensures controlled clamping pressure
  • Made from high-quality surgical stainless steel for durability and corrosion resistance
  • Autoclavable and reusable, suitable for repeated professional use
  • Ideal for microsurgery, ENT, ophthalmic use, pediatrics, and detailed procedural work

Specifications

Product Type Forceps
Material Stainless Steel
Style Straight
Tip Style Serrated
Length (English) 3.5 in.
Length (Metric) 8.9 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.