Learn More
Aviva Systems Biology Corp OR11H12 Antibody - N-terminal region (ARP44646_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44646_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OR11H12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human OR11H12. Alternate names: -ApplicationWB. Species reactivity: Cow, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-014-58863