Learn More
Aviva Systems Biology Corp OR13C5 Antibody - middle region (ARP31561_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31561_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OR13C5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human OR13C5. Alternate names: OR9-11ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rat
Catalog No. 50-014-54266